accessibility menu, dialog, popup

UserName

CAS RN 124858-35-1

CH3NHOFONOHO

CAS RN 124858-35-1

IUPAC Name: 7-fluoro-8-(4-hydroxypiperidin-1-yl)-12-methyl-4-oxo-1-azatricyclo[7.3.1.0⁵,¹³]trideca-2,5,7,9(13)-tetraene-3-carboxylic acid
Synonyms: nadifloxacin NDFX nadifloxacinum acuatim S-nadifloxacin S-ndfx R-ndfx nadifloxacino nadifloxacine
Molecular Weight (g/mol): 360.39
Molecular Formula: C19H21FN2O4
InChi Key: JYJTVFIEFKZWCJ-UHFFFAOYNA-N
SMILES: CC1CCC2=C3N1C=C(C(O)=O)C(=O)C3=CC(F)=C2N1CCC(O)CC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results