CAS RN 1246819-87-3
CAS RN 1246819-87-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Plerixafor-d4, TRC
CAS: 1246819-87-3 Molecular Formula: C28H50D4N8 Molecular Weight (g/mol): 506.81 Synonym: 1,1'-[1,4-(Phenylene-d4)bis(methylene)]bis-1,4,8,11-tetraazacyclotetradecane SMILES: [2H]C(C([2H])=C(CN1CCNCCCNCCNCCC1)C([2H])=C2[2H])=C2CN3CCCNCCNCCCNCC3