CAS RN 1246814-96-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
D-Beta-Imidazole lactic Acid Monohydrate, TRC
CAS: 1246814-96-9 Molecular Formula: C6 H8 N2 O3 . H2 O Molecular Weight (g/mol): 174.15 IUPAC Name: (2R)-2-hydroxy-3-(1H-imidazol-4-yl)propanoic acid;hydrate SMILES: O.O[C@H](Cc1c[nH]cn1)C(=O)O