accessibility menu, dialog, popup

UserName

H3CNOOOHO(E/Z)

CAS RN 124525-55-9

IUPAC Name: 2-[(3-nitrophenyl)methylidene]butanoic acid
Synonyms: 2-[(3-nitrophenyl)methylidene]butanoic acid
Molecular Weight (g/mol): 221.21
Molecular Formula: C11H11NO4
InChi Key: QMUHBYNLXQHZNR-UHFFFAOYSA-N
SMILES: CCC(=CC1=CC(=CC=C1)[N+]([O-])=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results