accessibility menu, dialog, popup

UserName

H2OH2OH2OH2OH2OOSOOOOV

CAS RN 12439-96-2

IUPAC Name: oxovanadiumbis(ylium) pentahydrate sulfate
Synonyms: oxovanadium(IV) pentahydrate sulfate
Molecular Weight (g/mol): 253.07
Molecular Formula: H10O10SV
InChi Key: YGYBXHQARYQUAY-UHFFFAOYSA-L
SMILES: O.O.O.O.O.O=[V++].[O-]S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)