accessibility menu, dialog, popup

UserName

CAS RN 1240257-82-2

IUPAC Name: 1-[3-chloro-5-(pentafluoro-λ⁶-sulfanyl)phenyl]methanamine
Synonyms: 1-[3-chloro-5-(pentafluoro-λ⁶-sulfanyl)phenyl]methanamine
Molecular Weight (g/mol): 267.64
Molecular Formula: C7H7ClF5NS
InChi Key: VUWNYLVTMIHGGJ-UHFFFAOYSA-N
SMILES: NCC1=CC(=CC(Cl)=C1)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results