accessibility menu, dialog, popup

UserName

SFFFFFFOH

CAS RN 1240257-69-5

IUPAC Name: [2-fluoro-4-(pentafluoro-λ⁶-sulfanyl)phenyl]methanol
Synonyms: [2-fluoro-4-(pentafluoro-λ⁶-sulfanyl)phenyl]methanol
Molecular Weight (g/mol): 252.17
Molecular Formula: C7H6F6OS
InChi Key: KOIOTESUJQFJAB-UHFFFAOYSA-N
SMILES: OCC1=C(F)C=C(C=C1)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results