accessibility menu, dialog, popup

UserName

OFFFFOHO

CAS RN 1240257-16-2

IUPAC Name: 3-[2-fluoro-4-(trifluoromethoxy)phenyl]propanoic acid
Synonyms: 3-[2-fluoro-4-(trifluoromethoxy)phenyl]propanoic acid
Molecular Weight (g/mol): 252.17
Molecular Formula: C10H8F4O3
InChi Key: XGMJOFWSONMRNZ-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=C(F)C=C(OC(F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results