CAS RN 1240257-03-7
CAS RN 1240257-03-7
IUPAC Name:
2-fluoro-4-(pentafluoro-λ⁶-sulfanyl)benzoic acid
Synonyms:
2-fluoro-4-(pentafluoro-λ⁶-sulfanyl)benzoic acid
Molecular Weight (g/mol):
266.16
Molecular Formula:
C7H4F6O2S
InChi Key:
MGIDEXWQQLSCGW-UHFFFAOYSA-N
SMILES:
OC(=O)C1=C(F)C=C(C=C1)S(F)(F)(F)(F)F
1
–
1
of
1
results
1
2-Fluoro-4-(pentafluorothio)benzoic acid, 97%, Thermo Scientific™
CAS: 1240257-03-7 Molecular Formula: C7H4F6O2S Molecular Weight (g/mol): 266.157 MDL Number: MFCD16652503 InChI Key: MGIDEXWQQLSCGW-UHFFFAOYSA-N Synonym: 2-fluoro-4-pentafluoro-??-sulfanyl benzoic acid,2-fluoro-4-pentafluorothio benzoic acid,2-fluoro-4-pentafluorosulfur benzoic acid PubChem CID: 66523491 IUPAC Name: 2-fluoro-4-(pentafluoro-$l^{6}-sulfanyl)benzoic acid SMILES: C1=CC(=C(C=C1S(F)(F)(F)(F)F)F)C(=O)O