accessibility menu, dialog, popup

UserName

CAS RN 1240256-92-1

IUPAC Name: 3-fluoro-5-(pentafluoro-λ⁶-sulfanyl)benzoyl chloride
Synonyms: 3-fluoro-5-(pentafluoro-λ⁶-sulfanyl)benzoyl chloride
Molecular Weight (g/mol): 284.6
Molecular Formula: C7H3ClF6OS
InChi Key: RCKDILSECAWCTQ-UHFFFAOYSA-N
SMILES: FC1=CC(=CC(=C1)C(Cl)=O)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results