accessibility menu, dialog, popup

UserName

CAS RN 124-19-6

H3CO

CAS RN 124-19-6

IUPAC Name: nonanal
Synonyms: nonanal pelargonaldehyde
Molecular Weight (g/mol): 142.24
Molecular Formula: C9H18O
InChi Key: GYHFUZHODSMOHU-UHFFFAOYSA-N
SMILES: CCCCCCCCC=O
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)