accessibility menu, dialog, popup

UserName

H3CCH3

CAS RN 124-18-5

IUPAC Name: decane
Synonyms: decane n-dekan decane (substance) dekan ccris 653 N-decane hsdb 63
Molecular Weight (g/mol): 142.29
Molecular Formula: C10H22
InChi Key: DIOQZVSQGTUSAI-UHFFFAOYSA-N
SMILES: CCCCCCCCCC
Molecular Weight (g/mol) 
  • (10)
  • (13)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (8)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (5)
Boiling Point 
  • (6)
  • (2)
  • (3)
  • (4)
  • (3)
Solvent or Matrix 
  • (2)
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (5)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (16)
  • (2)
  • (2)
  • (3)
  • (4)
  • (2)
  • (5)
  • (6)
  • (3)
  • (1)
  • (10)
  • (13)
  • (3)
  • (2)
  • (1)
  • (1)
  • (8)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (5)
  • (6)
  • (2)
  • (3)
  • (4)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (5)
  • (1)
  • (16)
  • (5)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (4)
  • (2)
  • (1)