accessibility menu, dialog, popup

UserName

CAS RN 124-03-8

BrH3CNH3CCH3CH3

CAS RN 124-03-8

IUPAC Name: ethyl(hexadecyl)dimethylazanium bromide
Synonyms: cetethyldimonium bromide
Molecular Weight (g/mol): 378.48
Molecular Formula: C20H44BrN
InChi Key: VUFOSBDICLTFMS-UHFFFAOYSA-M
SMILES: [Br-].CCCCCCCCCCCCCCCC[N+](C)(C)CC
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
pH 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results