accessibility menu, dialog, popup

UserName

NN

CAS RN 123847-85-8

IUPAC Name: N4,N4'-bis(naphthalen-1-yl)-N4,N4'-diphenyl-[1,1'-biphenyl]-4,4'-diamine
Synonyms: N4,N4'-bis(naphthalen-1-yl)-N4,N4'-diphenyl-[1,1'-biphenyl]-4,4'-diamine
Molecular Weight (g/mol): 588.75
Molecular Formula: C44H32N2
InChi Key: IBHBKWKFFTZAHE-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)N(C1=CC=C(C=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=C2C=CC=CC2=CC=C1)C1=C2C=CC=CC2=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results