accessibility menu, dialog, popup

UserName

CAS RN 123334-20-3

OSOOOOV

CAS RN 123334-20-3

IUPAC Name: oxovanadiumbis(ylium) hydrate sulfate
Synonyms: oxovanadium(IV) hydrate sulfate
Molecular Weight (g/mol): 181.01
Molecular Formula: H2O6SV
InChi Key: DKCWBFMZNUOFEM-UHFFFAOYSA-L
SMILES: O.O=[V++].[O-]S([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results