accessibility menu, dialog, popup

UserName

CAS RN 123333-98-2

CrFFF

CAS RN 123333-98-2

IUPAC Name: trifluorochromium hydrate
Synonyms: chromium trifluoride hydrate
Molecular Weight (g/mol): 127.01
Molecular Formula: CrF3H2O
InChi Key: BEUXKGJFVWBTKR-UHFFFAOYSA-K
SMILES: O.F[Cr](F)F
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results