accessibility menu, dialog, popup

UserName

CAS RN 123207-48-7

ONNH2

CAS RN 123207-48-7

IUPAC Name: 3-[(morpholin-4-yl)methyl]aniline
Synonyms: 3-(morpholin-4-ylmethyl)aniline
Molecular Weight (g/mol): 192.26
Molecular Formula: C11H16N2O
InChi Key: IHHKTIKNEDOMIK-UHFFFAOYSA-N
SMILES: NC1=CC=CC(CN2CCOCC2)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results