accessibility menu, dialog, popup

UserName

CAS RN 123-25-1

H3COOOOCH3

CAS RN 123-25-1

IUPAC Name: 1,4-diethyl butanedioate
Synonyms: diethyl succinate
Molecular Weight (g/mol): 174.2
Molecular Formula: C8H14O4
InChi Key: DKMROQRQHGEIOW-UHFFFAOYSA-N
SMILES: CCOC(=O)CCC(=O)OCC
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)