accessibility menu, dialog, popup

UserName

CAS RN 122955-13-9

ONNHClHCl

CAS RN 122955-13-9

IUPAC Name: 10,10-bis[(pyridin-4-yl)methyl]-9,10-dihydroanthracen-9-one dihydrochloride
Synonyms: 10,10-bis(pyridin-4-ylmethyl)anthracen-9-one dihydrochloride
Molecular Weight (g/mol): 449.38
Molecular Formula: C26H22Cl2N2O
InChi Key: WOGWMARIFDNZON-UHFFFAOYSA-N
SMILES: Cl.Cl.O=C1C2=CC=CC=C2C(CC2=CC=NC=C2)(CC2=CC=NC=C2)C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results