accessibility menu, dialog, popup

UserName

CAS RN 12277-88-2

IUPAC Name: platinum(2+) bis(benzenide) cycloocta-1,5-diene
Synonyms: platinum(2+) 1,5-cyclooctadiene bis(benzenide)
Molecular Weight (g/mol): 457.48
Molecular Formula: C20H22Pt
InChi Key: GGVUMOXWZYBTLY-UHFFFAOYSA-N
SMILES: [Pt++].C1=CC=[C-]C=C1.C1=CC=[C-]C=C1.C1CC=CCCC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results