accessibility menu, dialog, popup

UserName

OOBrF

CAS RN 1226808-75-8

IUPAC Name: 5-bromo-4-fluoro-2H-1,3-benzodioxole
Synonyms: 5-bromo-4-fluoro-2H-1,3-benzodioxole
Molecular Weight (g/mol): 219.01
Molecular Formula: C7H4BrFO2
InChi Key: FMEWVPPYMHXEGT-UHFFFAOYSA-N
SMILES: FC1=C(Br)C=CC2=C1OCO2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results