accessibility menu, dialog, popup

UserName

CAS RN 12257-42-0

RhRhClCl

CAS RN 12257-42-0

IUPAC Name: λ²-rhodium(2+) bis(bicyclo[2.2.1]hepta-2,5-diene) dichloride
Synonyms: λ²-rhodium(2+) bis(norbornadiene) dichloride
Molecular Weight (g/mol): 358.09
Molecular Formula: C14H16Cl2Rh
InChi Key: HCZNBZKRJTYNCE-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Rh++].C1C2C=CC1C=C2.C1C2C=CC1C=C2
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results