accessibility menu, dialog, popup

UserName

CAS RN 1225608-24-1

IUPAC Name: potassium (2,1,3-benzoxadiazol-5-yl)trifluoroboranuide
Synonyms: potassium 2,1,3-benzoxadiazol-5-yltrifluoroboranuide
Molecular Weight (g/mol): 226.01
Molecular Formula: C6H3BF3KN2O
InChi Key: VVXGHNBCIPLVLI-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)C1=CC2=NON=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results