accessibility menu, dialog, popup

UserName

ONOFFF

CAS RN 122454-46-0

IUPAC Name: 3-oxo-3-[4-(trifluoromethoxy)phenyl]propanenitrile
Synonyms: 3-oxo-3-[4-(trifluoromethoxy)phenyl]propanenitrile
Molecular Weight (g/mol): 229.16
Molecular Formula: C10H6F3NO2
InChi Key: IYZQGBBCANKWMX-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=C(C=C1)C(=O)CC#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results