CAS RN 122368-54-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-[(4-Chlorophenyl)(4-piperidinyloxy)methyl]pyridine, TRC
CAS: 122368-54-1 Molecular Formula: C17H19ClN2O Molecular Weight (g/mol): 302.8 Synonym: (RS)-4-[(4-Chlorophenyl)(2-pyridyl)methoxy]piperidine IUPAC Name: 2-[(4-chlorophenyl)-piperidin-4-yloxymethyl]pyridine SMILES: Clc1ccc(cc1)C(OC2CCNCC2)c3ccccn3