accessibility menu, dialog, popup

UserName

OO

CAS RN 122-60-1

IUPAC Name: 2-(phenoxymethyl)oxirane
Synonyms: phenyl glydidyl ether
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: FQYUMYWMJTYZTK-UHFFFAOYNA-N
SMILES: C(OC1=CC=CC=C1)C1CO1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)