accessibility menu, dialog, popup

UserName

CAS RN 1218790-49-8

BOOH3CCH3H3CH3COHNCH

CAS RN 1218790-49-8

IUPAC Name: N-(prop-2-yn-1-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
Synonyms: N-(prop-2-yn-1-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
Molecular Weight (g/mol): 285.15
Molecular Formula: C16H20BNO3
InChi Key: BUZIZNWTLHGQQO-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C(=O)NCC#C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results