accessibility menu, dialog, popup

UserName

CAS RN 121846-11-5

H3COOOOSiCH3H3CH3CCH3H3CH3C

CAS RN 121846-11-5

IUPAC Name: tert-butyl({2,2-dimethyl-2H,4H,4aH,8H,8aH-pyrano[3,2-d][1,3]dioxin-8-yl}oxy)dimethylsilane
Synonyms: tert-butyl({2,2-dimethyl-4H,4aH,8H,8aH-pyrano[3,2-d][1,3]dioxin-8-yl}oxy)dimethylsilane
Molecular Weight (g/mol): 300.47
Molecular Formula: C15H28O4Si
InChi Key: PYHWHKQRALSXOY-UHFFFAOYNA-N
SMILES: CC(C)(C)[Si](C)(C)OC1C=COC2COC(C)(C)OC12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results