CAS RN 1217226-46-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Nitrofurantoin 13C3, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
Nitrofurantoin-13C3, TRC
CAS: 1217226-46-4 Molecular Formula: 13C3 C5 H6 N4 O5 Molecular Weight (g/mol): 241.14 Synonym: Labeled Nitrofurantoin,Nitrofurantoin-13C3 IUPAC Name: 1-[(E)-(5-nitrofuran-2-yl)methylideneamino]-(2,4,5-^{13}C_{3})1,3-diazolidine-2,4-dione SMILES: [O-][N+](=O)c1oc(\C=N\N2[13CH2][13C](=O)N[13C]2=O)cc1