CAS RN 1216850-33-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Desethyldesisopropyl Atrazine-13C3, TRC
CAS: 1216850-33-7 Molecular Formula: 13C3 H4 Cl N5 Molecular Weight (g/mol): 148.53 Synonym: Atrazine-desethyl-desisopropyl 13C3,6-Chloro-2,4-diamino-1,3,5-triazine 13C3 (2,4,6-13C3),Atrazine-desethyl-desisopropyl 13C3 (2,4,6-13C3) IUPAC Name: 6-chloro-(2,4,6-^{13}C_{3})1,3,5-triazine-2,4-diamine SMILES: N[13c]1n[13c](N)n[13c](Cl)n1
2
Honeywell Fluka™ 6-Chloro-2,4-diamino-1,3,5-triazine-13C3
Analytical standard, reference material