CAS RN 1216459-54-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ibuprofen-13C6, TRC
CAS: 1216459-54-9 Molecular Formula: C713C6H18O2 Molecular Weight (g/mol): 212.24 Synonym: 2-(4-Isobutylphenyl)propionic Acid-13C6; SMILES: CC(C(O)=O)[13C]1=[13CH][13CH]=[13C](CC(C)C)[13CH]=[13CH]1