accessibility menu, dialog, popup

UserName

CAS RN 1215564-15-0

NNFFFNNSOOCl

CAS RN 1215564-15-0

IUPAC Name: 1-[4-(trifluoromethyl)pyrimidin-2-yl]-1H-pyrazole-4-sulfonyl chloride
Synonyms: 1-[4-(trifluoromethyl)pyrimidin-2-yl]pyrazole-4-sulfonyl chloride
Molecular Weight (g/mol): 312.65
Molecular Formula: C8H4ClF3N4O2S
InChi Key: SADUDBJMXGGXDE-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=NC(=NC=C1)N1C=C(C=N1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results