accessibility menu, dialog, popup

UserName

CAS RN 12129-69-0

IUPAC Name: 1,3,5-trimethylbenzene tris(methanidylidyneoxidanium) tungsten
Synonyms: tris(carbon monoxide) mesitylene tungsten
Molecular Weight (g/mol): 388.07
Molecular Formula: C12H12O3W
InChi Key: OENLQXWEAUYNBC-UHFFFAOYSA-N
SMILES: [W].[C-]#[O+].[C-]#[O+].[C-]#[O+].CC1=CC(C)=CC(C)=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results