accessibility menu, dialog, popup

UserName

CAS RN 1212734-83-2

HNFFF

CAS RN 1212734-83-2

IUPAC Name: 2-(trifluoromethyl)piperidine
Synonyms: 2-(trifluoromethyl)piperidine
Molecular Weight (g/mol): 153.15
Molecular Formula: C6H10F3N
InChi Key: NAXDEFXCCITWEU-UHFFFAOYNA-N
SMILES: FC(F)(F)C1CCCCN1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results