accessibility menu, dialog, popup

UserName

CAS RN 12125-08-5

NH4NH4ClClClClClClOs

CAS RN 12125-08-5

IUPAC Name: osmium(4+) diammonium hexachloride
Synonyms: osmium(4+) diammonium hexachloride
Molecular Weight (g/mol): 439.01
Molecular Formula: Cl6H8N2Os
InChi Key: SRBXXQDKBKTWOC-UHFFFAOYSA-J
SMILES: [NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Os+4]
Physical Form 
  • (2)
  • (4)
Quantity 
  • (1)
  • (3)
  • (2)
Grade 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (2)
  • (4)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (4)