accessibility menu, dialog, popup

UserName

CAS RN 1211866-85-1

CH3HNOCH3NHNNNNOOFFF

CAS RN 1211866-85-1

IUPAC Name: N-(2-methyl-3-{[4-(4-{[4-(trifluoromethoxy)phenyl]methoxy}piperidin-1-yl)-1,3,5-triazin-2-yl]amino}phenyl)acetamide
Synonyms: N-(2-methyl-3-{[4-(4-{[4-(trifluoromethoxy)phenyl]methoxy}piperidin-1-yl)-1,3,5-triazin-2-yl]amino}phenyl)acetamide
Molecular Weight (g/mol): 516.53
Molecular Formula: C25H27F3N6O3
InChi Key: QLKAFHZJICDACE-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=CC(NC2=NC=NC(=N2)N2CCC(CC2)OCC2=CC=C(OC(F)(F)F)C=C2)=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results