CAS RN 121104-96-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Celgosivir, TRC
CAS: 121104-96-9 Molecular Formula: C12H21NO5 Molecular Weight (g/mol): 259.3 Synonym: MDL 28574,6-Butyryl castanospermine,Castanospermine 6-butyrate IUPAC Name: [(1S,6S,7S,8R,8aR)-1,7,8-trihydroxy-1,2,3,5,6,7,8,8a-octahydroindolizin-6-yl] butanoate SMILES: O=C(OCC(OC(NC1=CC=CC=C1)=O)CN2CCCCC2)NC3=CC=CC=C3.Cl