accessibility menu, dialog, popup

UserName

O

CAS RN 1210-35-1

IUPAC Name: tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-one
Synonyms: tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-one
Molecular Weight (g/mol): 208.26
Molecular Formula: C15H12O
InChi Key: BMVWCPGVLSILMU-UHFFFAOYSA-N
SMILES: O=C1C2=CC=CC=C2CCC2=CC=CC=C12
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)