CAS RN 121-43-7
Trimethyl borate, 99.99% (metals basis)
CAS: 121-43-7 Molecular Formula: C3H9BO3 Molecular Weight (g/mol): 103.912 MDL Number: MFCD00008346 InChI Key: WRECIMRULFAWHA-UHFFFAOYSA-N Synonym: trimethoxyborane,methyl borate,trimethoxyboron,boric acid, trimethyl ester,trimethylborate,boric acid trimethyl ester,trimethoxyborine,borester o,boric acid h3bo3 , trimethyl ester,b och3 3 PubChem CID: 8470 ChEBI: CHEBI:38913 IUPAC Name: trimethyl borate SMILES: B(OC)(OC)OC
Trimethyl borate, 99%, AcroSeal™
CAS: 121-43-7 Molecular Formula: C3H9O3B Molecular Weight (g/mol): 103.91 MDL Number: MFCD00008346 InChI Key: WRECIMRULFAWHA-UHFFFAOYSA-N Synonym: trimethoxyborane,methyl borate,trimethoxyboron,boric acid, trimethyl ester,trimethylborate,boric acid trimethyl ester,trimethoxyborine,borester o,boric acid h3bo3 , trimethyl ester,b och3 3 PubChem CID: 8470 ChEBI: CHEBI:38913 IUPAC Name: trimethyl borate SMILES: B(OC)(OC)OC
Trimethyl borate, 99%, Thermo Scientific Chemicals
CAS: 121-43-7 Molecular Formula: C3H9BO3 Molecular Weight (g/mol): 103.912 MDL Number: MFCD00008346 InChI Key: WRECIMRULFAWHA-UHFFFAOYSA-N Synonym: trimethoxyborane,methyl borate,trimethoxyboron,boric acid, trimethyl ester,trimethylborate,boric acid trimethyl ester,trimethoxyborine,borester o,boric acid h3bo3 , trimethyl ester,b och3 3 PubChem CID: 8470 ChEBI: CHEBI:38913 IUPAC Name: trimethyl borate SMILES: B(OC)(OC)OC
Trimethyl borate, 99%
CAS: 121-43-7 MDL Number: MFCD00008346 InChI Key: WRECIMRULFAWHA-UHFFFAOYSA-N Synonym: trimethoxyborane,methyl borate,trimethoxyboron,boric acid, trimethyl ester,trimethylborate,boric acid trimethyl ester,trimethoxyborine,borester o,boric acid h3bo3 , trimethyl ester,b och3 3 PubChem CID: 8470 ChEBI: CHEBI:38913 IUPAC Name: trimethyl borate SMILES: B(OC)(OC)OC
Trimethyl borate, 99.9995+% (metals basis)
CAS: 121-43-7 Molecular Formula: C3H9BO3 Molecular Weight (g/mol): 103.912 MDL Number: MFCD00008346 InChI Key: WRECIMRULFAWHA-UHFFFAOYSA-N Synonym: trimethoxyborane,methyl borate,trimethoxyboron,boric acid, trimethyl ester,trimethylborate,boric acid trimethyl ester,trimethoxyborine,borester o,boric acid h3bo3 , trimethyl ester,b och3 3 PubChem CID: 8470 ChEBI: CHEBI:38913 IUPAC Name: trimethyl borate SMILES: B(OC)(OC)OC
Trimethyl Borate, Sigma-Aldrich™
CAS: 121-43-7