accessibility menu, dialog, popup

UserName

CAS RN 12091-64-4

MoMoCOCOCOCOCOCO

CAS RN 12091-64-4

IUPAC Name: [1,1-bis(oxidaniumylidynemethanuidyl)-1-[1,1,1-tris(oxidaniumylidynemethanuidyl)-1-molybdapentacyclo[2.2.0.0¹,³.0¹,⁵.0²,⁶]hexa-2,4-dien-1-yl]-1-molybdapentacyclo[2.2.0.0¹,³.0¹,⁵.0²,⁶]hexa-2,4-dien-1-yl](oxidaniumylidyne)methanuide
Synonyms: [1,1-bis(oxidaniumylidynemethanuidyl)-1-[1,1,1-tris(oxidaniumylidynemethanuidyl)-1-molybdapentacyclo[2.2.0.0¹,³.0¹,⁵.0²,⁶]hexa-2,4-dien-1-yl]-1-molybdapentacyclo[2.2.0.0¹,³.0¹,⁵.0²,⁶]hexa-2,4-dien-1-yl](oxidaniumylidyne)methanuide
Molecular Weight (g/mol): 482.09
Molecular Formula: C16H2Mo2O6
InChi Key: AYBJYGWNVNZERR-UHFFFAOYSA-N
SMILES: C1=2C3=C4[Mo]131([Mo]3567(C8=C3C5C6=C78)([C-]#[O+])([C-]#[O+])[C-]#[O+])([C-]#[O+])([C-]#[O+])([C-]#[O+])C4C=21
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results