accessibility menu, dialog, popup

UserName

MoMo

CAS RN 12069-89-5

IUPAC Name: methane dimolybdenum
Synonyms: methane dimolybdenum
Molecular Weight (g/mol): 207.94
Molecular Formula: CH4Mo2
InChi Key: JAGQSESDQXCFCH-UHFFFAOYSA-N
SMILES: C.[Mo].[Mo]
Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results