accessibility menu, dialog, popup

UserName

CAS RN 1206641-26-0

BOOH3CCH3H3CH3CNSOOCH3

CAS RN 1206641-26-0

IUPAC Name: 3-methanesulfonyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Synonyms: 3-methanesulfonyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Weight (g/mol): 283.15
Molecular Formula: C12H18BNO4S
InChi Key: BFWBUXKEBXAKDE-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(=CN=C1)S(C)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results