CAS RN 12046-04-7
CAS RN 12046-04-7
IUPAC Name:
nonaammonium tetrahydrate pentaborate
Synonyms:
nonaammonium tetrahydrate pentaborate
Molecular Weight (g/mol):
528.45
Molecular Formula:
B5H44N9O19
InChi Key:
OJAJAQRDJBKPPU-UHFFFAOYSA-W
SMILES:
[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].O.O.O.O.[O-]B([O-])[O-].[O-]B([O-])[O-].[O-]B([O-])[O-].[O-]B([O-])[O-].[O-]B([O-])[O-]
1
–
1
of
1
results
1
Ammonium pentaborate tetrahydrate, 98%
CAS: 12046-04-7 Molecular Formula: B5H44N9O19-6 Molecular Weight (g/mol): 528.446 MDL Number: MFCD13176621 InChI Key: OJAJAQRDJBKPPU-UHFFFAOYSA-W Synonym: nonaammonium tetrahydrate pentaborate PubChem CID: 57347064 IUPAC Name: nonaazanium;pentaborate;tetrahydrate SMILES: B([O-])([O-])[O-].B([O-])([O-])[O-].B([O-])([O-])[O-].B([O-])([O-])[O-].B([O-])([O-])[O-].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].O.O.O.O