accessibility menu, dialog, popup

UserName

TbTbOOOTbTbOOOO

CAS RN 12037-01-3

IUPAC Name: diterbium(3+) trioxidandiide
Synonyms: terbium(III) oxide
Molecular Weight (g/mol): 365.85
Molecular Formula: O3Tb2
InChi Key: SCRZPWWVSXWCMC-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[Tb+3].[Tb+3]
Physical Form 
  • (7)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (4)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (13)
  • (1)
  • (7)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)
  • (3)