accessibility menu, dialog, popup

UserName

OOOTmTm

CAS RN 12036-44-1

IUPAC Name: dithulium(3+) trioxidandiide
Synonyms: thulium(III) oxide
Molecular Weight (g/mol): 385.87
Molecular Formula: O3Tm2
InChi Key: ZIKATJAYWZUJPY-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[Tm+3].[Tm+3]
Physical Form 
  • (7)
  • (2)
Quantity 
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (7)
  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)