accessibility menu, dialog, popup

UserName

CAS RN 12036-39-4

IUPAC Name: strontium(2+) oxozirconiumbis(olate)
Synonyms: strontium(2+) oxozirconiumbis(olate)
Molecular Weight (g/mol): 226.84
Molecular Formula: O3SrZr
InChi Key: FCCTVDGKMTZSPU-UHFFFAOYSA-N
SMILES: [Sr++].[O-][Zr]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results