accessibility menu, dialog, popup

UserName

CAS RN 12030-49-8

IrOO

CAS RN 12030-49-8

IUPAC Name: iridium tetrahydroxide
Synonyms: iridium(IV) oxide
Molecular Weight (g/mol): 260.25
Molecular Formula: H4IrO4
InChi Key: UOYXEZMXOVJLIH-UHFFFAOYSA-J
SMILES: [OH-].[OH-].[OH-].[OH-].[Ir]
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results