accessibility menu, dialog, popup

UserName

CAS RN 12007-01-1

BNiNi

CAS RN 12007-01-1

IUPAC Name: boron dinickel
Synonyms: nickel boride
Molecular Weight (g/mol): 128.2
Molecular Formula: BNi2
InChi Key: WRLJWIVBUPYRTE-UHFFFAOYSA-N
SMILES: [B].[Ni].[Ni]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results