accessibility menu, dialog, popup

UserName

ZnZnAsZnAs

CAS RN 12006-40-5

IUPAC Name: bis(zincio-λ²-arsanyl) zinc
Synonyms: bis(zincio-λ²-arsanyl) zinc
Molecular Weight (g/mol): 345.98
Molecular Formula: As2Zn3
InChi Key: RHKSESDHCKYTHI-UHFFFAOYSA-N
SMILES: [Zn].[Zn]=[As].[Zn]=[As]
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results