accessibility menu, dialog, popup

UserName

CAS RN 120-73-0

HNNNN

CAS RN 120-73-0

IUPAC Name: 7H-purine
Synonyms: purine
Molecular Weight (g/mol): 120.12
Molecular Formula: C5H4N4
InChi Key: KDCGOANMDULRCW-UHFFFAOYSA-N
SMILES: N1C=NC2=NC=NC=C12
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)